missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiregulin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16036-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype.Spezifikation
Amphiregulin | |
Polyclonal | |
Unconjugated | |
AREG | |
amphiregulin, AR, AREG, AREGB, CRDGF, SDGF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
374 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15514 | |
AREG | |
Amphiregulin Fusion Protein Ag8907 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |