Learn More
Anacardic acid, Tocris Bioscience™
Noncompetitive PCAF/p300 inhibitor
Marke:  RD Systems (Bio-Techne) 3084/10
Weitere Details : Netto-Gewicht : 0.00010kg
Menge. 10mg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13412364
  • 132.30 € / 10 Milligramm
Voraussichtlicher Liefertermin 27-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Noncompetitive inhibitor of PCAF and p300 histone acetyltransferase (HAT) activity (IC 50 values are ∽5.0 and ∽8.5 μM respectively). Inhibits NF-κB activation. Also reported to selectively activate Aurora kinase A-mediated phosphorylation of histone H3 and to inhibit protein SUMOylation. Exhibits antitumor, antimicrobial and antioxidant activity.
Spezifikation
Chemische Identifikatoren
16611-84-0
348.527
anacardic acid, 6-pentadecylsalicylic acid, cyclogallipharic acid, 22:0-anacardic acid, 2-hydroxy-6-pentadecyl-benzoic acid, ginkgolic acid c15:0, anacardsaure, anarcadic acid, anacardic acid a, anacardic acid /
CHEBI:2696
CCCCCCCCCCCCCCCC1=C(C(=CC=C1)O)C(=O)O
C22H36O3
ADFWQBGTDJIESE-UHFFFAOYSA-N
167551
2-hydroxy-6-pentadecylbenzoic acid
Spezifikation
Anacardic acid
16611-84-0
anacardic acid, 6-pentadecylsalicylic acid, cyclogallipharic acid, 22:0-anacardic acid, 2-hydroxy-6-pentadecyl-benzoic acid, ginkgolic acid c15:0, anacardsaure, anarcadic acid, anacardic acid a, anacardic acid /
CCCCCCCCCCCCCCCC1=C(C(=CC=C1)O)C(=O)O
348.527
CHEBI:2696
>97%
10mg
C22H36O3
ADFWQBGTDJIESE-UHFFFAOYSA-N
2-hydroxy-6-pentadecylbenzoic acid
167551
348.52
Videos