missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-like 3 Antibody, R&D Systems™
Rat Monoclonal Antibody
N.V.
Spezifikation
Antigen | Angiopoietin-like Protein 3/ANGPTL3 |
---|---|
Klon | 128611 |
Konzentration | LYOPH |
Verdünnung | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Anwendungen | ELISA |
Beschreibung
Angiopoietin-like Protein 3/ANGPTL3 Monoclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Spezifikation
Angiopoietin-like Protein 3/ANGPTL3 | |
LYOPH | |
ELISA | |
Unconjugated | |
Rat | |
Mouse | |
Q9R182 | |
27329 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse Angiopoietin-like 3 Ser17-Thr455 Accession # Q9R182 | |
Primary | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Detects mouse Angiopoietin-like 3 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human ANGPTL3, recombinant mouse (rm) ANGPTL4, or rmAngiopoietin-3 is observed. |
128611 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
ANGPTL3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only