missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiostatin Kringle 1-3 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
N.V.
Spezifikation
Antigen | Angiostatin |
---|---|
Klon | 79735 |
Verdünnung | Western Blot 1 ug/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Beschreibung
Angiostatin Monoclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.Spezifikation
Angiostatin | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
5340 | |
E. coli-derived recombinant human Angiostatin Kringle 1-3 Val98-Ser358 (Cys88Ser, Cys316Ser) Accession # P00747 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Angiostatin Kringle 1-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody also detects plasminogen but does not cross-react with recombinant mouse Angiostatin. |
79735 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P00747 | |
PLG | |
IgG2a | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only