Beschreibung
ANKRD6 Polyclonal specifically detects ANKRD6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Spezifikation
Spezifikation
| Antigen | ANKRD6 |
| Anwendungen | Immunocytochemistry, Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Verdünnung | Immunocytochemistry/Immunofluorescence 1-4 ug/ml |
| Zusammensetzung | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Gen-Alias | ankyrin repeat domain 6, diversin, KIAA0957ankyrin repeat domain-containing protein 6 |
| Gensymbole | ANKRD6 |
| Wirtsspezies | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KDDRRRKSRPKVSAFSDPTPPADQQPGHQKNLHAHNHPKKRNRHRCSSPPPPHEFRAYQLYTLYRGKDGKVMQAPINGCRCEPLINKLE |
| Mehr anzeigen |