missing translation for 'onlineSavingsMsg'
Learn More

Annexin IV Mouse anti-Human, Clone: 1A10B5, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 60288-2-IG-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887355

  • 112.71 € / 20 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
Spezifikation

Spezifikation

Annexin IV
1A10B5
Unconjugated
ANXA4
Liquid
ANXA4
Annexin IV Fusion Protein Ag0130
Protein A
RUO
Antibody
Monoclonal
Human
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09525
35-beta calcimedin, Annexin A4, Annexin IV, Annexin-4, Chromobindin-4, Endonexin I, Lipocortin IV
Mouse
IgG2c
20 μL
-20°C
Primary
307
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation