missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 Mouse, Unlabeled, Clone: 12/14-3-3, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 561465

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15881049

  • 304.38 € / 1 Stück
Verfügbar ab: 19-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The family of 14-3-3 proteins has been implicated in numerous cellular processes such as neurotransmission, phospholipase activity, and calcium signaling. The 14-3-3 protein can associate with several proteins involved in cell proliferation, such as Raf-1, Bcr-Abl, middle T antigen, and CDC25 phosphatase. It appears that 14-3-3 proteins enhance the kinase activity of the Raf proto-oncogene equally as well as Ras, thus, implicating 14-3-3 proteins in the Raf/Ras/Map kinase signaling pathway. However, in spite of 14-3-3 direct association with the CDC25 phosphatase, this interaction does not increase the phosphatase activity. It has been reported that this antibody detects 14-3-3 subunits β, γ, η, Θ, and ζ , but not ε nor σ.

Spezifikation

Spezifikation

14-3-3
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human 14-3-3
50 μg
Primary
Store undiluted at -20°C.
Western Blot, Immunohistochemistry, Immunofluorescence
12/14-3-3
Unconjugated
Mouse
Affinity Purified
RUO
Rat, Mouse, Human, Frog, Chicken, Canine
IgM
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.