missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Monoclonal Antibody (HIS.H8), Alexa Fluor™ 647, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA121315A647
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for Western Blot.Spezifikation
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Mouse | |
Protein A | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Western Blot | |
HIS.H8 | |
Alexa Fluor 647 | |
6His tag; 6X His; His Tag; Histidine Tag | |
6x His synthetic peptide | |
50 μL | |
Primary | |
Antibody | |
IgG2b |