missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Monoclonal Antibody (4A12E4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen 372900
Weitere Details : Netto-Gewicht : 0.02100kg
Beschreibung
6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
6x-His Tag | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
-20°C | |
Liquid | |
Tag |
Immunocytochemistry, Western Blot | |
4A12E4 | |
Unconjugated | |
6His tag; 6X His; His Tag; Histidine Tag | |
Synthetic peptide containing six His residues (6xHis). | |
100 μg | |
Primary | |
Antibody | |
IgG1 κ |