missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Monoclonal Antibody (4E3D10H2/E3), HRP, Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA1135HRP
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
MA1-135-HRP detects both N-terminal and C-terminal His-tagged proteins in Western Blot applications.
6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for Western Blot.Spezifikation
6x-His Tag | |
Monoclonal | |
1 mg/mL | |
PBS with proprietary stabilizer and 0.07% Kathon | |
Mouse | |
Protein A | |
RUO | |
4° C, do not freeze | |
Liquid | |
Tag |
Western Blot | |
4E3D10H2/E3 | |
HRP | |
6His tag; 6X His; His Tag; Histidine Tag | |
Mixture of N-terminal and C-terminal 6x-His peptides, CG-HHHHHH and HHHHHH-GC | |
50 μL | |
Primary | |
Antibody | |
IgG1 |