missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A33 Monoclonal Antibody (402104), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA524139
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse A33 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
A33 Monoclonal antibody specifically detects A33 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Western Blot.Spezifikation
A33 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q99795 | |
Gpa33 | |
Mouse myeloma cell line NS0-derived recombinant human A33 Ile22-Val235 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry, Western Blot | |
402104 | |
Unconjugated | |
Gpa33 | |
2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33 | |
Rat | |
Protein A/G | |
RUO | |
10223 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |