missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A33 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA547444
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse A33 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
A33 Polyclonal antibody specifically detects A33 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
A33 | |
Polyclonal | |
Unconjugated | |
Gpa33 | |
2010310L10Rik; 2210401D16Rik; A33; A33 antigen; BB116197; cell surface A33 antigen; Glycoprotein A33; glycoprotein A33 (transmembrane); GPA33; mA33; MGC129986; MGC129987; transmembrane glycoprotein A33 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10223 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q99795 | |
Gpa33 | |
Mouse myeloma cell line NS0-derived recombinant human A33 Ile22-Val235 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |