missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A4GALT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA523625
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A4GALT Polyclonal antibody specifically detects A4GALT in Human samples. It is validated for Western Blot.Spezifikation
A4GALT | |
Polyclonal | |
Unconjugated | |
A4galt | |
A14GALT; A4GALT; A4galt {ECO:0000312; A4GALT1; alpha 1,4-galactosyltransferase; alpha 14-galactosyltransferase; alpha-1,4-galactosyltransferase; Alpha-1,4-N-acetylglucosaminyltransferase; alpha4Gal-T1; cd77; CD77 synthase; Gb3; Gb3 synthase; Gb3/CD77 synthase; Gb3S; Globotriaosylceramide synthase; lac; lactosylceramide 4-alpha-galactosyltransferase; P blood group (P one antigen); P(k); P(k) antigen synthase; P1; P1/Pk synthase; P1PK; PK; RGD:621583}; truncated alpha 1,4-galactosyltransferase; UDP-galactose:beta-D-galactosyl-beta1-R 4-alpha-D-galactosyltransferase | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
53947 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.28 mg/mL | |
PBS with 0.09% sodium azide | |
Q9NPC4 | |
A4galt | |
KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human A4GALT | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |