missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AADAT Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518859
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
AADAT Polyclonal antibody specifically detects AADAT in Human samples. It is validated for Western Blot.Spezifikation
AADAT | |
Polyclonal | |
Unconjugated | |
Aadat | |
2-aminoadipate aminotransferase; 2-aminoadipate transaminase; AadAT; Aadt; AI875679; alpha-aminoadipate aminotransferase; aminoadipate aminotransferase; KAT/AadAT; Kat2; KATII; KYAT2; kynurenine aminotransferase 2; kynurenine aminotransferase II; kynurenine/alpha-aminoadipate aminotransferase, mitochondrial; kynurenine--oxoglutarate; Kynurenine--oxoglutarate aminotransferase II; kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate transaminase II; L kynurenine/alpha aminoadipate aminotransferase; mKat-2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
51166 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8N5Z0 | |
Aadat | |
Synthetic peptide sequence (KPEDAKNPQKNTPK) corresponding to the internal amino acids of AADAT | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |