missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AARSD1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519203
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine and canine based on sequence homology.
AARSD1 Polyclonal antibody specifically detects AARSD1 in Human samples. It is validated for Western Blot.Spezifikation
AARSD1 | |
Polyclonal | |
Unconjugated | |
AARSD1 | |
1110069E20Rik; 2310044P18Rik; AA589600; AARSD1; Alanyl-tRNA deacylase alaX; alanyl-tRNA editing protein Aarsd1; alanyl-tRNA synthetase domain containing 1; alanyl-tRNA synthetase domain-containing protein 1; Alax; AlaXp-II; MGC2744 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
80755 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BTE6 | |
AARSD1 | |
Synthetic peptide sequence (IEFYAKVNSKDSQDK) corresponding to the internal amino acids of AARSD1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |