missing translation for 'onlineSavingsMsg'
Learn More

alpha-1 Antitrypsin Monoclonal Antibody (2B12), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Thermo Scientific MA515521

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11354403

  • 530.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA5-15521 targets AAT in indirect ELISA, WB applications and shows reactivity with Human and mouse samples. The MA5-15521 immunogen is purified recombinant fragment of human AAT expressed in E. Coli. MA5-15521 detects AAT which has a predicted molecular weight of approximately 47kDa.

alpha-1 Antitrypsin Monoclonal antibody specifically detects alpha-1 Antitrypsin in Human, Mouse samples. It is validated for ELISA, Western Blot.
Spezifikation

Spezifikation

alpha-1 Antitrypsin
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
P01009, P07758
SERPINA1, Serpina1a
Purified recombinant fragment of human AAT expressed in E. Coli.
RUO
20700, 5265
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
2B12
Unconjugated
SERPINA1
a-1 AT; A1A; a1-antitrypsin; A1AT; AAT; Aat2; Aat-2; alpha 1 antitrypsin; alpha-1 antitrypsin; alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha1AT; alpha-1-protease inhibitor; alpha-1-proteinase inhibitor; Digestive Zymogen; Dom1; MGC23330; MGC9222; PI; PI1; PRO0684; PRO2209; PRO2275; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; protease inhibitor 1 (anti-elastase, antitrypsin, alpha-1); Protease Serine 1; serine (or cysteine) peptidase inhibitor, clade A, member 1A; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A (alpha-1antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Proteas
Mouse
100 μL
Primary
Human, Mouse
Antibody
IgG1
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote