missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AATK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546795
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: APNRPQQADG SPNGSTAEEG GGFAWDDDFP LMTAKAAFAM ALDPAAPAPA
AATK Polyclonal antibody specifically detects AATK in Human samples. It is validated for Western Blot.Spezifikation
AATK | |
Polyclonal | |
Unconjugated | |
AATK | |
Aatk; AATYK; aatyk1; apoptosis associated tyrosine kinase; apoptosis-associated tyrosine kinase; apoptosis-associated tyrosine kinase 1B; brain apoptosis-associated tyrosine kinase; CDK5-binding protein; KIAA0641; Lemur tyrosine kinase 1; LMR1; LMTK1; mKIAA0641; p35-binding protein; p35BP; PPP1R77; protein phosphatase 1, regulatory subunit 77; serine/threonine-protein kinase LMTK1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9625 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZMQ8 | |
AATK | |
Synthetic peptide directed towards the C-terminal of human AATK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |