missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB5 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518844
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ABCB5 Polyclonal antibody specifically detects ABCB5 in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
ABCB5 | |
Polyclonal | |
Unconjugated | |
ABCB5 | |
9230106F14Rik; ABCB5; ABCB5 P-gp; ABCB5alpha; ABCB5beta; ATP binding cassette subfamily B member 5; ATP-binding cassette protein; ATP-binding cassette sub-family B member 5; ATP-binding cassette, subfamily B (MDR/TAP), member 5; ATP-binding cassette, sub-family B (MDR/TAP), member 5; EST422562; P-glycoprotein ABCB5; RGD1566342 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
340273 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q2M3G0 | |
ABCB5 | |
Synthetic peptide sequence (QTQHRNTSKKAQ ) corresponding to the internal amino acids of ABCB5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |