missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518315
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse based on sequence homology.
MRP4 Polyclonal antibody specifically detects MRP4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
MRP4 | |
Polyclonal | |
Unconjugated | |
ABCC4 | |
ABCC4; ABC-tranporter; ATP binding cassette subfamily C member 4; ATP-binding cassette protein C4; ATP-binding cassette sub-family C member 4; ATP-binding cassette, subfamily C (CFTR/MRP), member 4; ATP-binding cassette, sub-family C (CFTR/MRP), member 4; bA464I2.1 (ATP-binding cassette, sub-family C (CFTR/MRP), member 4); canalicular multispecific organic anion transporter (ABC superfamily); D630049P08Rik; EST170205; MOATB; MOAT-B; MRP/cMOAT-related ABC transporter; MRP4; Multidrug resistance-associated protein 4; multi-specific organic anion transporter B; RP11-74A12.1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10257 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O15439 | |
ABCC4 | |
Synthetic peptide sequence (CNGQPSTLTIFETAL) corresponding to the C-terminus amino acids of ABCC4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |