missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRP6 Monoclonal Antibody (M6II-7), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA135685
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MRP6 Monoclonal antibody specifically detects MRP6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.Spezifikation
MRP6 | |
Monoclonal | |
250 μg/mL | |
tissue culture supernatant with 1% BSA and 0.1% sodium azide | |
O95255 | |
ABCC6 | |
ABCC6. | |
RUO | |
368 | |
4° C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
M6II-7 | |
Unconjugated | |
ABCC6 | |
ABC34; Abcc1b; ABCC6; Anthracycline resistance-associated protein; ARA; ATP binding cassette subfamily C member 6; ATP-binding cassette sub-family C member 6; ATP-binding cassette, subfamily C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C (CFTR/MRP), member 6; ATP-binding cassette, sub-family C, member 6; DCC; dyscalc; Dyscalc1; EST349056; GACI2; liver multidrug resistance-associated protein 6; Mlp1; MLP-1; MOATE; MOAT-E; Mrp6; MRP-like protein 1; Multidrug resistance-associated protein 6; multidrug resistance-associated protein-6; multi-specific organic anion transporter E; PXE; PXE1; URG7; URG7 protein; multidrug resistance-associated protein 6 | |
Rat | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG2a |