missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCD1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518967
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ABCD1 Polyclonal antibody specifically detects ABCD1 in Human samples. It is validated for Western Blot.Spezifikation
ABCD1 | |
Polyclonal | |
Unconjugated | |
ABCD1 | |
ABC42; ABCD1; Adrenoleukodystrophy protein; ALD; Aldgh; ALDP; AMN; ATP binding cassette subfamily D member 1; ATP-binding cassette sub-family D member 1; ATP-binding cassette, subfamily D (ALD), member 1; ATP-binding cassette, sub-family D (ALD), member 1; ATP-binding cassette, sub-family D, member 1; RGD1562128; X-linked adrenoleukodystrophy (ALD) gene homolog | |
Goat | |
Antigen affinity chromatography | |
RUO | |
215 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P33897 | |
ABCD1 | |
Synthetic peptide sequence (EDMQRKGYSEQD) corresponding to the internal amino acids of ABCD1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |