missing translation for 'onlineSavingsMsg'
Learn More

ABCD2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA542397

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15575432

  • 472.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Peptide sequence: WRFEQLDTAI RLTLSEEKQK LESQLAGIPK MQQRLNELCK ILGEDSVLKT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 85%; Zebrafish: 79%

ABCD2 Polyclonal antibody specifically detects ABCD2 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

ABCD2
Polyclonal
Unconjugated
ABCD2
ABC39; Abcd2; adrenoleukodystrophy related; adrenoleukodystrophy-like 1; Adrenoleukodystrophy-related protein; ALD1; ALDL1; ALDR; ALDRP; ATP binding cassette subfamily D member 2; ATP-binding cassette sub-family D member 2; ATP-binding cassette, subfamily D (ALD), member 2; ATP-binding cassette, sub-family D (ALD), member 2; ATP-binding cassette, sub-family D, member 2; hALDR; LOW QUALITY PROTEIN: ATP-binding cassette sub-family D member 2
Rabbit
Affinity Chromatography
RUO
225
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UBJ2
ABCD2
Synthetic peptide directed towards the middle region of human ABCD2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote