Learn More
ABCG8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA116792
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody does not react with mouse samples and is not recommended for IHC applications.
ABCG8 Polyclonal antibody specifically detects ABCG8 in Human samples. It is validated for Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Spezifikation
ABCG8 | |
Polyclonal | |
Unconjugated | |
Abcg8 | |
1300003C16Rik; ABCG8; AI114946; ATP binding cassette subfamily G member 8; ATP-binding cassette sub-family G member 8; ATP-binding cassette, subfamily G (WHITE), member 8; ATP-binding cassette, sub-family G (WHITE), member 8; ATP-binding cassette, subfamily G, member 8; GBD4; MGC142217; sterolin 2; Sterolin2; sterolin-2; STSL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64241 | |
4° C, do not freeze | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H221 | |
Abcg8 | |
A synthetic peptide from the N-terminal region of human ABCG8 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |