missing translation for 'onlineSavingsMsg'
Learn More

ABHD9 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA539001

Weitere Details : Netto-Gewicht : 0.00900kg

Artikelnummer. 15555911

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 06-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The alpha/ beta hydrolase superfamily comprise diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical alpha/ beta hydrolase fold, which consists of an eightstranded parallel alpha/ beta structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer′s disease, obesity and blood clotting disorders. The abhydrolase domain-containing protein 9 (ABHD9), also designated epoxide hydrolase 3 (EPHX3), is a 360 amino acid protein and a member of the alpha/ beta hydrolase superfamily. The gene encoding ABHD9 maps to chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.
Spezifikation

Spezifikation

ABHD9
Polyclonal
Unconjugated
EPHX3
Epoxide hydrolase 3, Abhydrolase domain-containing protein 9, EPHX3, ABHD9
Rabbit
Antigen affinity chromatography
RUO
79852
-20°C
Liquid
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9H6B9
EPHX3
A synthetic peptide derived from the internal region of human ABHD9
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote