missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABHD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536418
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 41 kDa. Purity is >95% by SDS-PAGE.
ABHD9 Polyclonal antibody specifically detects ABHD9 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ABHD9 | |
Polyclonal | |
Unconjugated | |
EPHX3 | |
2310063B19Rik; Abhd9; abhydrolase domain containing 9; abhydrolase domain-containing protein 9; EH3; Ephx3; Epoxide hydrolase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
366836, 71932, 79852 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3V1F8, Q9H6B9 | |
EPHX3 | |
Synthetic peptide corresponding to amino acids 110-157 of human ABHD9 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |