Learn More
ABI3BP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531867
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-31867 targets ABI3BP in WB applications and shows reactivity with Human samples. The PA5-31867 immunogen is recombit protein fragment corresponding to a region within amino acids 704 and 903 of Human ABI3BP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Spezifikation
ABI3BP | |
Polyclonal | |
Unconjugated | |
Q7Z7G0 | |
ABI3BP | |
Recombit protein fragment corresponding to a region within amino acids 704 and 903 of Human ABI3BP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.76 mg/mL | |
ABI3BP | |
NESHBP, TARSH, ABI gene family member 3-binding protein, ABI gene family, member 3 (NESH) binding protein, nesh-binding protein, target of Nesh-SH3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25890 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |