missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Abl Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA514761
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with mouse based on sequence homology.
c-Abl Polyclonal antibody specifically detects c-Abl in Human samples. It is validated for Western Blot.Spezifikation
c-Abl | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P00519 | |
ABL1 | |
KLH conjugated synthetic peptide between 231-260 amino acids from the N-terminal region of human ABL1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ABL1 | |
Abelson murine leukemia oncogene; Abelson murine leukemia viral (v-abl) oncogene homolog 1; abelson murine leukemia viral oncogene homolog 1; Abelson tyrosine-protein kinase 1; Abl; Abl 1 antib; ABL proto-oncogene 1, non-receptor tyrosine kinase; Abl1; ABL2; ABLL; AI325092; ARG; B30; bcr/abl; bcr/c-abl oncogene protein; c-ABL; c-abl oncogene 1, non-receptor tyrosine kinase; c-abl oncogene 1, receptor tyrosine kinase; c-ABL1; E430008G22Rik; JTK7; p150; Proto-oncogene c-Abl; proto-oncogene tyrosine-protein kinase ABL1; RP11-83J21.1; Tyrosine kinase ARG; tyrosine-protein kinase ABL1; v-abl; v-abl Abelson murine leukemia oncogene 1; v-abl Abelson murine leukemia viral oncogene 1; v-abl Abelson murine leukemia viral oncogene homolog 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
25 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |