Learn More
Abl2 Monoclonal Antibody (1H1B11), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515330
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15330 targets ABL2 in indirect ELISA, ICC and WB applications and shows reactivity with Human samples. The MA5-15330 immunogen is purified recombinant fragment of ABL2 expressed in E. Coli.
Abl2 Monoclonal antibody specifically detects Abl2 in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
Abl2 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P42684 | |
ABL2 | |
Purified recombinant fragment of ABL2 expressed in E. Coli. | |
RUO | |
27 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1H1B11 | |
Unconjugated | |
ABL2 | |
AA536808; Abelson murine leukemia viral (v-abl) oncogene homolog 2; abelson murine leukemia viral oncogene homolog 2; Abelson tyrosine-protein kinase 2; Abelson-related gene protein; ABL proto-oncogene 2, non-receptor tyrosine kinase; Abl1-related gene product; abl2; ABLL; ARG; arg tyrosine kinase; c-abl oncogene 2, non-receptor tyrosine kinase; FLJ22224; FLJ31718; FLJ41441; tyrosine kinase ARG; tyrosine-protein kinase ABL2; Tyrosine-protein kinase ARG; v-abl Abelson murine leukemia viral oncogene 2; v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene); v-abl Abelson murine leukemia viral oncogene homolog 2; v-abl Abelson murine leukemia viral oncogene homolog 2 (arg, Abelson-related gene) | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |