Learn More
ABT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540569
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: EVAQAKRETD FYLQSVERGQ RFLAADGDPA RPDGSWTFAQ RPTEQELRAR Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%; Zebrafish: 77%
ABT1 Polyclonal antibody specifically detects ABT1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ABT1 | |
Polyclonal | |
Unconjugated | |
Abt1 | |
ABT1; Activator of basal transcription 1; basal transcriptional activator; hABT1; hypothetical protein LOC553568; im:7139827; TATA-binding protein-binding protein; zgc:109995 | |
Rabbit | |
Protein A | |
RUO | |
29777 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9ULW3 | |
Abt1 | |
Synthetic peptide directed towards the middle region of human ABT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |