missing translation for 'onlineSavingsMsg'
Learn More

ACBD3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA544816

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15596642

  • 472.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Peptide sequence: EARRLEQRWG FGLEELYGLA LRFFKEKDGK AFHPTYEEKL KLVALHKQVL Sequence homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%

ACBD3 Polyclonal antibody specifically detects ACBD3 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

ACBD3
Polyclonal
Unconjugated
Acbd3
60kDa; 8430407O11Rik; Acbd3; acyl-CoA binding domain containing 3; acyl-CoA-binding domain-containing protein 3; acyl-Coenzyme A binding domain containing 3; D1Ertd10e; DAP; DMT1-associated protein; fa20g08; GCP60; Gocap1; golgi complex associated protein 1, 60 kDa; golgi complex associated protein 1, 60kDa; golgi complex-associated protein 1; Golgi phosphoprotein 1; Golgi resident protein GCP60; Golgi resident protein GCP60, N-terminally processed; GOLPH1; neurotrophin receptor alike death domain protein; p75-like apoptosis-inducing death domain protein; Pap7; PBR- and PKA-associated protein 7; PBR associated protein; Peripheral benzodiazepine receptor-associated protein PAP7; peripherial benzodiazepine receptor associated protein; PKA (RIalpha)-associated protein; si:dkey-267p15.1; wu:fa20g08; zgc:66303
Rabbit
Affinity Chromatography
RUO
64746
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9H3P7
Acbd3
Synthetic peptide directed towards the N-terminal of human ACBD3
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote