Learn More
ACBD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543439
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: NSLGKMSREE AMSAYITEMK LVAQKVIDTV PLGEVAEDMF GYFEPLYQVI Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
ACBD4 Polyclonal antibody specifically detects ACBD4 in Human samples. It is validated for Western Blot.Spezifikation
ACBD4 | |
Polyclonal | |
Unconjugated | |
ACBD4 | |
2010009P05Rik; 2010015A21Rik; Acbd4; acyl-CoA binding domain containing 4; acyl-CoA-binding domain-containing protein 4; acyl-Coenzyme A binding domain containing 4; AI849317; FLJ13322; FLJ90623; HMFT0700; similar to acyl-Coenzyme A binding domain containing 4; zgc:85611 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79777 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NC06 | |
ACBD4 | |
Synthetic peptide directed towards the N-terminal of human ACBD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |