Learn More
ASIC4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530649
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: SK-N-SH, U87-MG, MCF-7. Predicted reactivity: Mouse (96%), Rat (96%), Rabbit (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ASIC4 Polyclonal antibody specifically detects ASIC4 in Human samples. It is validated for Western Blot.Spezifikation
ASIC4 | |
Polyclonal | |
Unconjugated | |
Asic4 | |
Accn4; acid sensing (proton gated) ion channel family member 4; acid sensing ion channel family member 4; acid sensing ion channel subunit family member 4; acid-sensing (proton-gated) ion channel family member 4; acid-sensing ion channel 4; Amiloride-sensitive cation channel 4; Amiloride-sensitive cation channel 4, pituitary; amiloride-sensitive cation channel family member 4, pituitary; ASIC4; BNAC4; brain sodium channel 4; MGC17248; MGC24860; putative acid-sensing ion channel; Spasic; Spinal chord ASIC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55515 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.42 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96FT7 | |
Asic4 | |
Recombinant fragment corresponding to a region within amino acids 85 and 323 of Human ACCN4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |