Learn More
ACAT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA124413
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-24413 detects Acetyl-CoA Acetyltransferase (ACAT2) in human samples. PA1-24413 has been successfully used in Western blot and immunohistochemistry (paraffin) procedures. The PA1-24413 immunogen is a synthetic peptide corresponding to a region of human ACAT2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Acetyl-Coenzyme A acetyltransferase 2 is an enzyme involved in lipid metabolism. Reported patients with ACAT2 deficiency have shown severe mental retardation and hypotonus. The ACAT2 gene shows complementary overlapping with the 3-prime region of the TCP1 gene in both mouse and human. These genes are encoded on opposite strands of DNA, as well as in opposite transcriptional orientation.Spezifikation
ACAT2 | |
Polyclonal | |
Unconjugated | |
Q9BWD1 | |
ACAT2 | |
Synthetic peptide corresponding to a region of human ACAT2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
ACAT2 | |
ACAT2 | |
Rabbit | |
Protein A | |
RUO | |
39 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |