Learn More
H2BK15ac Recombinant Rabbit Monoclonal Antibody (7H17L5), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen 701799
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Since it is highly conserved across species, the antibody may react with many other species. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
H2BK15ac Recombinant Monoclonal antibody specifically detects H2BK15ac in Human samples. It is validated for Immunocytochemistry, Peptide Array, Western Blot.Spezifikation
H2BK15ac | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P62807 | |
HIST1H2BC | |
Acetylated peptide (Lys15) corresponding to Human H2 (aa 11-18) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Peptide Array, Western Blot | |
7H17L5 | |
Unconjugated | |
HIST1H2BC | |
2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K15; Acetyl-Histone H2B Lys15; AV127319; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2B 291A; H2B 291B; H2b 613; H2B GL105; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member S; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; | |
Rabbit | |
Protein A | |
RUO | |
8347 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |