missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Phosphatase 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141703
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-41703 detects Acid phosphatase 1, isoform b. Suggested positive control: antigen standard for ACP1 (transient overexpression lysate), human liver protein, liver lysate.
Acid Phosphatase 1 Polyclonal antibody specifically detects Acid Phosphatase 1 in Bovine, Human, Porcine samples. It is validated for Western Blot.Spezifikation
Acid Phosphatase 1 | |
Polyclonal | |
Unconjugated | |
ACP1 | |
4632432E04Rik; acid phosphatase; acid phosphatase 1; acid phosphatase 1, soluble; acid phosphatase of erythrocyte; acp1; Acp-1; Acp1-PA; adipocyte acid phosphatase; Adult cuticle protein 1; AI427468; CG7216; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; HAAP betaÊ; hypothetical protein; LMW-PTP; LMW-PTPase; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; Low molecular weight cytosolic acid phosphatase; low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; RCJMB04_11a4; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37; Unknown (protein for MGC:128334) | |
Rabbit | |
Protein G | |
RUO | |
100737301, 280977, 52 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
P11064, P24666, P81693 | |
ACP1 | |
Synthetic peptide corresponding to residues 40-80 of human ACP1. | |
100 μg | |
Primary | |
Bovine, Human, Porcine | |
Antibody | |
IgG |