missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acinus Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA146272
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Acinus Polyclonal antibody specifically detects Acinus in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Acinus | |
Polyclonal | |
Unconjugated | |
ACIN1 | |
2610036I19Rik; 2610510L13Rik; Acin1; ACINUS; acinusL; acinusS; Acn; apoptotic chromatin condensation inducer 1; apoptotic chromatin condensation inducer in the nucleus; C79325; DKFZp667N107; fSAP152; functional spliceosome-associated protein 152; KIAA0670; mKIAA0670 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9UKV3 | |
ACIN1 | |
C-terminal sequence RNRQLEREKRREHS. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |