missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aconitase 2 Monoclonal Antibody (7G4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA1029X
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Aconitase 2 Monoclonal antibody specifically detects Aconitase 2 in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Spezifikation
Aconitase 2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q99798, Q99KI0, Q9ER34 | |
Aco2 | |
Full length human recombinant protein of human ACO2 (NP_001089) produced in HEK293T cell. | |
20 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
7G4 | |
Unconjugated | |
Aco2 | |
Aco2; Aco-2; Aco3; Aconitase; aconitase 2; aconitase 2, mitochondrial; aconitase precursor; aconitate hydratase, mitochondrial; ACONM; citrate hydro-lyase; D10Wsu183e; epididymis secretory sperm binding protein Li 284; heart aconitase; heart aconitase precursor; HEL-S-284; hypothetical protein; ICRD; MGC20605; MGC33908; mitochondrial aconitase; mitochondrial aconitase (nuclear aco2 gene); OCA8; OPA9; RP3-347H13.8 | |
Mouse | |
Protein A | |
RUO | |
11429, 50, 79250 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |