missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA512212
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ACOX1 Polyclonal antibody specifically detects ACOX1 in Human samples. It is validated for Western Blot.Spezifikation
ACOX1 | |
Polyclonal | |
Unconjugated | |
ACOX1 | |
Acox; Acox1; acyl-CoA oxidase 1; acyl-CoA oxidase 1, palmitoyl; acyl-CoA oxidase, straight-chain; acyl-Coenzyme A oxidase 1, palmitoyl; AOX; D130055E20Rik; PALMCOX; palmitoyl-CoA oxidase; Paox; Peroxisomal acyl-CoA oxidase 1, A chain; Peroxisomal acyl-CoA oxidase 1, B chain; Peroxisomal acyl-CoA oxidase 1, C chain; Peroxisomal acyl-coenzyme A oxidase 1; peroxisomal fatty acyl-CoA oxidase; peroxisomal fatty acyl-coenzyme A oxidase; peroxisomal straight-chain Acyl-CoA oxidase; RATACOA1; SCOX; Straight-chain acyl-CoA oxidase | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
51 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q15067 | |
ACOX1 | |
KLH conjugated synthetic peptide between 629-659 amino acids from the C-terminal region of human ACOX1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |