missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOX2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518670
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with rabbit based on sequence homology.
ACOX2 Polyclonal antibody specifically detects ACOX2 in Human samples. It is validated for Western Blot.Spezifikation
ACOX2 | |
Polyclonal | |
Unconjugated | |
ACOX2 | |
3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA 24-hydroxylase; 3-alpha,7-alpha,12-alpha-trihydroxy-5-beta-cholestanoyl-CoA oxidase; Acox2; acyl-CoA oxidase 2; acyl-CoA oxidase 2, branched chain; acyl-Coenzyme A oxidase 2, branched chain; BCOX; branched chain acyl-CoA oxidase; BRCACOX; BRCOX; Peroxisomal acyl-coenzyme A oxidase 2; peroxisomal branched chain acyl-CoA oxidase; THCA-CoA oxidase; THCCox; trihydroxycoprostanoyl-CoA oxidase | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8309 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q99424 | |
ACOX2 | |
Synthetic peptide sequence (QWAQKSPTNTQENP) corresponding to the internal amino acids of ACOX2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |