missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACOX3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522373
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ACOX3 Polyclonal antibody specifically detects ACOX3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACOX3 | |
Polyclonal | |
Unconjugated | |
ACOX3 | |
Acox3; acyl-CoA oxidase 3, pristanoyl; acyl-Coenzyme A oxidase 3, pristanoyl; BI685180; branched-chain acyl-CoA oxidase; BRCACox; BRCOX; EST-s59; PCOX; Peroxisomal acyl-coenzyme A oxidase 3; PRCOX; Pristanoyl-CoA oxidase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80911, 8310 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O15254, Q9EPL9 | |
ACOX3 | |
Recombinant protein fragment contain a sequence corresponding to a region within amino acids 408 and 603 (O15254) of ACOX3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |