Learn More
NDUFAB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA522191
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-22191 targets ACP in ICC, IF, and WB applications and shows reactivity with Human, mouse and rat samples. The PA5-22191 immunogen is a synthetic peptide corresponding to a region within amino acids 93 and 156 of NDUFAB1 (Uniprot IDNo.O14561). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Spezifikation
NDUFAB1 | |
Polyclonal | |
Unconjugated | |
O14561, Q9CR21 | |
NDUFAB1 | |
Synthetic peptide corresponding to a region within amino acids 93 and 156 of NDUFAB1 (Uniprot IDNo.O14561) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
NDUFAB1 | |
ACP, FASN2A, SDAP, CI-SDAP, NADH-ubiquinone oxidoreductase 9.6 kDa subunit, NADH:ubiquinone oxidoreductase SDAP subunit, acyl carrier protein, mitochondrial, complex I SDAP subunit, mitochondrial acyl carrier protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
293453, 4706, 70316 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |