Learn More
Acid Phosphatase 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529961
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, HeLa, HepG2, H3347, CX-1, SW48, HCT116, Human Lymphoma Tissue. Predicted reactivity: Mouse (87%), Rat (87%), Pig (94%), Chimpanzee (100%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acid Phosphatase 2 Polyclonal antibody specifically detects Acid Phosphatase 2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Acid Phosphatase 2 | |
Polyclonal | |
Unconjugated | |
Acp2 | |
acid phosphatase 2, lysosomal; Acid phosphatase 2, lysozymal; Acp2; Acp-2; LAP; LMW-PTP-II low molecular weight phosphotyrosine protein phosphatase isozyme 2; lysosomal acid phosphatase; Unknown (protein for MGC:142461) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P11117 | |
Acp2 | |
Recombinant fragment corresponding to a region within amino acids 126 and 360 of Human ACP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |