missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA512225
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ACSL4 Polyclonal antibody specifically detects ACSL4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACSL4 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O60488 | |
Acsl4 | |
KLH conjugated synthetic peptide between 236-267 amino acids from the central region of human ACSL4 (FACL4) | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Acsl4 | |
9430020A05Rik; Acs4; ACSL4; acyl-CoA synthetase 4; acyl-CoA synthetase long-chain family member 4; Arachidonate--CoA ligase; AU018108; Facl4; fatty acid Coenzyme A ligase, long chain 4; fatty acid-Coenzyme A ligase long chain 4; fatty acid-Coenzyme A ligase, long chain 4; fatty-acid-Coenzyme A ligase, long-chain 4; L MRX63; LACS 4; Lacs4; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 4; long-chain fatty-acid-Coenzyme A ligase 4; long-chain-fatty-acid--CoA ligase 4; mACS4; MRX63; MRX68 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2182 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |