Learn More
Activin A Precursor Mouse anti-Human, Clone: 132815, R&D Systems™
Mouse Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1203
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418445
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 31-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Activin A Precursor Monoclonal antibody MAB1203 Specifically detects Activin A Precursor in Human samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Activin A
132815
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
INHBA
DNA construct encoding human Activin A pro region fused to mature human Activin beta B chain
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects human Activin A Precursor in direct ELISAs and Western blots. Recognizes an epitope found in the human Activin A pro region that is removed when the protein is processed to its mature form.
Human
Western Blot
LYOPH
Western Blot 1 ug/mL
Purified
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.