Learn More
ACVR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528662
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: mouse heart, rat liver. Predicted reactivity: Mouse (99%), Rat (99%), Xenopus laevis (93%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ACVR1 Polyclonal antibody specifically detects ACVR1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Spezifikation
ACVR1 | |
Polyclonal | |
Unconjugated | |
ACVR1 | |
activin A receptor type 1; activin A receptor type I; activin A receptor, type 1; activin A receptor, type I; activin A receptor, type II-like kinase 2; activin receptor I precursor; activin receptor type I; Activin receptor type-1; activin receptor-like kinase 2; activin type I receptor; ACTRI; ActR-I; ActRIA; Acvr; ACVR1; ACVR1A; ACVRLK2; ALK2; Alk-2; ALK2-B; Alk8; ATVR1; D330013D15Rik; FOP; hydroxyalkyl-protein kinase; serine/threonine-protein kinase receptor; serine/threonine-protein kinase receptor R1; SKR1; TGF-B superfamily receptor type I; Tgfb1; Tsk7L; TSK-7L; TSRI; TSR-I; type I TGF B receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11477, 79558, 90 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
0.9 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P37172, P80201, Q04771 | |
ACVR1 | |
Recombinant fragment corresponding to a region within amino acids 214 and 509 of Human Activin A receptor type I | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |