Learn More
ACVRL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527477
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ACVRL1 Polyclonal antibody specifically detects ACVRL1 in Human samples. It is validated for Western Blot.Spezifikation
ACVRL1 | |
Polyclonal | |
Unconjugated | |
ACVRL1 | |
activin A receptor like type 1; activin A receptor type II-like 1; activin A receptor type IL; activin A receptor, type II-like 1; activin A receptor, type II-like kinase 1; Activin receptor like kinase 1; activin receptor-like kinase 1; activin receptor-like kinase-1; Acvrl1; Acvrlk1; AI115505; AI427544; ALK1; ALK-1; HHT; HHT2; ORW2; R3; R-3; Serine/threonine-protein kinase receptor R3; SETHKIR; SKR3; TGF-B superfamily receptor type I; TSR-I | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P37023 | |
ACVRL1 | |
Recombinant fragment corresponding to a region within amino acids 311 and 503 of Human Activin A Receptor Type II-like 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |