Learn More
ACVR2A Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA120174
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: muscle fibroblasts. Neutralization: when used at a 1-3 μg/mL, the antibody will block 50% of the binding of 30 ng/mL recombinant human activin A to immobilized recombinant human activin receptor IIA/Fc chimera (100 μL of a 0.5 μg/mL solution coating each well) in a functional ELISA assay.
ACVR2A Polyclonal antibody specifically detects ACVR2A in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
ACVR2A | |
Polyclonal | |
Unconjugated | |
ACVR2A | |
activin A receptor type 2A; activin A receptor type IIA; activin A receptor, type IIA; activin receptor IIA; activin receptor type IIA; Activin receptor type-2A; Actrii; ActrIIa; ACTR-IIA; ACVR2; Acvr2a; rActR-II; TactrII; type II activin receptor | |
Goat | |
Antigen affinity chromatography | |
RUO | |
92 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 5% trehalose and no preservative | |
P27037 | |
ACVR2A | |
Recombinant fragment of human Activin Receptor Type IIA. | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |