Learn More
Actin-like 7A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530411
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (91%), Rat (92%), Chimpanzee (98%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Actin-like 7A Polyclonal antibody specifically detects Actin-like 7A in Human samples. It is validated for Western Blot.Spezifikation
Actin-like 7A | |
Polyclonal | |
Unconjugated | |
ACTL7A | |
actin like 7A; actin-like 7a; actin-like 7-alpha; actin-like protein 7A; actin-like Tact2; actin-like-7-alpha; ACTL7A; Arp7a; Tact2; T-actin 2; T-actin-2; testicular secretory protein Li 3; testis-specific actin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10881 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y615 | |
ACTL7A | |
Recombinant fragment corresponding to a region within amino acids 226 and 435 of Human ACTL7A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |