Learn More
AGK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528566
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A431, H1299, mouse heart. Predicted reactivity: Mouse (97%), Rat (96%), Pig (95%), Rhesus Monkey (99%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
AGK Polyclonal antibody specifically detects AGK in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
AGK | |
Polyclonal | |
Unconjugated | |
AGK | |
2610037M15Rik; 6720408I04Rik; acylglycerol kinase; acylglycerol kinase, mitochondrial; Agk; AI465370; CATC5; CTRCT38; FLJ10842; hAGK; hsMuLK; MTDPS10; Mulk; multiple substrate lipid kinase; multi-substrate lipid kinase; RGD1562046 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55750, 69923 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.84 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q53H12, Q9ESW4 | |
AGK | |
Recombinant fragment corresponding to a region within amino acids 1 and 235 of Human Acylglycerol kinase | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |