missing translation for 'onlineSavingsMsg'
Learn More

Adaptin ε Mouse anti-Human, Unlabeled, Clone: 32, BD

Mouse Monoclonal Antibody

Marke:  BD Biosciences 612018

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15896069

  • 308.94 € / 1 Set
Verfügbar ab: 19-12-2022
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Sorting of integral membrane proteins is mediated by vesicular trafficking between a variety of organelles. Two sorting signals are tyrosine-based and dileucine-based signals that interact with heterotetrameric adaptor protein complexes (AP-1, AP-2, AP-3, and AP-4), which are associated with the vesicle coats. These coatomers contain two large adaptin proteins (γ, α, σ, ε and β1, β2, βσ2, σ3, σ4 respectively). The AP-1 and AP-3 complexes are involved in protein sorting from the TGN and endosomes, while AP-2 adaptor complexes are involved in clathrin-mediated endocytosis. AP-4 is associated with non-clathrin coated vesicles in the region of the TGN. This localization is disrupted by brefeldin A, indicating that AP-4 membrane attachment is regulated by small GTPases. The µ4 subunit of the AP-4 complex interacts with tyrosine-based signals on LAMP-2 during targeting to the endosomal-lysosomal system. Thus, AP-4 is a less abundant AP complex that may be important for vesicle trafficking from the Golgi to the endosomal-lysosomal system.

Spezifikation

Spezifikation

Adaptin ε
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Adaptin ε aa. 685-793
50 μg
Cell Biology
Human
IgG1
Western Blot
32
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote

For Research Use Only.